Assay Details
Target Gene Details
Entrez Gene ID: | 56433 |
Gene Name: | vacuolar protein sorting 29 (S. pombe) |
Gene Aliases: |
2010015D08Rik, AW049835, PEP11 |
Location: |
Chr.5:122354377-122364981 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps29 | NM_019780.1 | 4 | 891 | NP_062754.1 |
| AB578901.1 | 3 | 663 | ||
| AF193794.1 | 4 | 891 | AAF04595.1 | |
| AK008248.1 | 5 | 911 | ||
| AK036426.1 | 4 | 881 | ||
| AK150330.1 | 4 | 901 | BAE29472.1 | |
| BC005663.1 | 4 | 880 | AAH05663.1 |
More Information
Set Membership: |
|

Genomic Map