Assay Details
Target Gene Details
Entrez Gene ID: | 319352 |
Gene Name: | PILR alpha associated neural protein |
Gene Aliases: |
AI255183, AU067746, C530028O21Rik, Panp |
Location: |
Chr.6:124996720-125003096 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pianp | NM_001145926.1 | 6 | 1694 | NP_001139398.1 |
| NM_001145927.1 | 6 | 1678 | NP_001139399.1 | |
| NM_175696.4 | 6 | 1849 | NP_783627.2 | |
| AK082984.1 | 6 | 1851 | BAC38721.1 | |
| AK208418.1 | ||||
| AK211709.1 | 1 | 250 | ||
| AK216678.1 | 1 | 226 | ||
| AK219084.1 | 1 | 210 | ||
| BC065170.1 | 6 | 1678 | AAH65170.1 |

Genomic Map