Assay Details
Target Gene Details
Entrez Gene ID: | 14789 |
Gene Name: | prolyl 3-hydroxylase 3 |
Gene Aliases: |
BC016431, Grcb, Leprel2 |
Location: |
Chr.6:124841095-124857746 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P3h3 | NM_013534.4 | 15 | 2281 | NP_038562.2 |
| AJ441086.1 | 15 | 2281 | CAD29580.1 | |
| AK140962.1 | 14 | 2515 | ||
| BC003726.1 | 15 | 1867 | AAH03726.1 | |
| BC016431.1 | 14 | 1767 | AAH16431.1 |
More Information
Set Membership: |
|

Genomic Map