Assay Details
Target Gene Details
Entrez Gene ID: | 114643 |
Gene Name: | 2'-5' oligoadenylate synthetase 1C |
Gene Aliases: |
Oasl2, Oasl5 |
Location: |
Chr.5:120800194-120812540 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oas1c | NM_033541.4 | 2 | 398 | NP_291019.1 |
| AB067528.1 | 1 | 55 | BAB62296.1 | |
| AF459815.1 | 1 | 55 | AAM47560.1 | |
| AF459816.1 | 1 | 55 | AAM47561.1 | |
| AK028986.1 | 2 | 399 | BAC26225.1 | |
| AK078952.1 | 1 | 66 | BAC37475.1 | |
| AK163253.1 | 1 | 71 | BAE37260.1 | |
| BC108955.1 | 1 | 64 | AAI08956.1 | |
| BC126877.1 | 1 | 63 |
More Information
Set Membership: |
|

Genomic Map