Assay Details
Target Gene Details
Entrez Gene ID: | 353172 |
Gene Name: | glycyl-tRNA synthetase |
Gene Aliases: |
GENA202, Gena201, Nmf249, Sgrp23 |
Location: |
Chr.6:55038001-55079504 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gars | NM_180678.3 | 17 | 2201 | NP_851009.2 |
| AK012758.2 | 17 | 2196 | ||
| AK146238.1 | 17 | 2178 | BAE27003.1 | |
| AK165857.1 | 17 | 2149 | BAE38417.1 | |
| AK190866.1 | 1 | 72 | ||
| AK191772.1 | 2 | 242 | ||
| AK192609.1 | 1 | 72 | ||
| AK193995.1 | 1 | 72 | ||
| AK198633.1 | 1 | 72 | ||
| AK202425.1 | 1 | 72 | ||
| AK203383.1 | 1 | 96 | ||
| AK205044.1 | 1 | 72 | ||
| AV236862.1 | 1 | 48 | ||
| BC021747.1 | 17 | 2182 | AAH21747.1 |
More Information
Set Membership: |
|

Genomic Map