Assay Details
Target Gene Details
Entrez Gene ID: | 14562 |
Gene Name: | growth differentiation factor 3 |
Gene Aliases: |
C78318, Gdf-3, Vgr-2, Vgr2, ecat9 |
Location: |
Chr.6:122605403-122610071 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdf3 | NM_008108.4 | 1 | 254 | NP_032134.2 |
| AK082877.1 | 1 | 174 | ||
| AK146174.1 | 1 | 180 | BAE26953.1 | |
| AK152457.1 | 1 | 255 | BAE31235.1 | |
| AK160533.1 | 1 | 212 | BAE35850.1 | |
| AK167962.1 | 1 | 180 | BAE39959.1 | |
| BC101963.1 | 1 | 169 | AAI01964.1 | |
| BC101964.1 | 1 | 169 | AAI01965.1 | |
| BC103565.1 | 1 | 168 | AAI03566.1 | |
| L06443.1 | 1 | 270 | AAA53034.1 | |
| S52658.1 | 1 | 210 | AAB24876.1 |
More Information
Set Membership: |
|

Genomic Map