Assay Details
Target Gene Details
Entrez Gene ID: | 15199 |
Gene Name: | heme binding protein 1 |
Gene Aliases: |
Hebp |
Location: |
Chr.6:135137519-135168215 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hebp1 | NM_013546.2 | 4 | 699 | NP_038574.2 |
| AB013095.1 | 4 | 638 | BAA33770.1 | |
| AF117613.1 | 4 | 687 | AAD32096.1 | |
| AK008919.1 | 4 | 620 | ||
| BC012654.1 | 4 | 623 | AAH12654.1 | |
| BE692297.1 | 1 | 357 | ||
| CT010262.1 | CAJ18470.1 |

Genomic Map