Assay Details
Target Gene Details
Entrez Gene ID: | 23983 |
Gene Name: | poly(rC) binding protein 1 |
Gene Aliases: |
WBP17, [a]CP-1, alphaCP-1 |
Location: |
Chr.6:86524494-86526171 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcbp1 | NM_011865.3 | 1 | 295 | NP_035995.1 |
| AF073936.1 | 1 | 328 | ||
| AF139894.1 | 1 | 64 | AAD51920.1 | |
| AK002433.1 | 1 | 54 | ||
| BC004793.1 | 1 | 156 | AAH04793.1 | |
| BC069915.1 | 1 | 295 | AAH69915.1 |
More Information
Set Membership: |
|

Genomic Map