Assay Details
Target Gene Details
Entrez Gene ID: | 78412 |
Gene Name: | RIKEN cDNA 3110062M04 gene |
Gene Aliases: |
AL023049, Mri |
Location: |
Chr.6:34871771-34879019 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110062M04Rik | NM_001135611.1 | 4 | 2239 | NP_001129083.1 |
| NM_199145.2 | 4 | 2564 | NP_954596.1 | |
| AK042380.1 | 4 | 2520 | BAC31243.1 | |
| AK083365.1 | 4 | 2206 |
Target Gene Details
Entrez Gene ID: | 68487 |
Gene Name: | transmembrane protein 140 |
Gene Aliases: |
1110007F12Rik |
Location: |
Chr.6:34863106-34874946 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem140 | NM_197986.2 | 2 | 600 | NP_932103.1 |
| AK030532.1 | 2 | 601 | BAC27007.1 | |
| BC020080.1 | 1 | 413 | AAH20080.1 | |
| BC023802.1 | 2 | 614 | AAH23802.1 |

Genomic Map