Assay Details
Target Gene Details
Entrez Gene ID: | 100535 |
Gene Name: | 2'-5' oligoadenylate synthetase 1D |
Gene Aliases: |
AA536864, C85127, oasl8 |
Location: |
Chr.5:120914818-120921648 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oas1d | NM_133893.3 | 2 | 345 | NP_598654.1 |
| AB067532.1 | 2 | 299 | BAB84132.1 | |
| AK054465.1 | 2 | 313 | BAC35788.1 | |
| AK136001.1 | 2 | 313 | BAE22768.1 | |
| AY055829.1 | 2 | 307 | AAL12825.1 | |
| BC052835.1 | 2 | 309 | AAH52835.1 | |
| DV659301.1 | 2 | 345 |
More Information
Set Membership: |
|

Genomic Map