Assay Details
Target Gene Details
Entrez Gene ID: | 24116 |
Gene Name: | negative elongation factor complex member A, Whsc2 |
Gene Aliases: |
Whsc2, Whsc2h |
Location: |
Chr.5:33898179-33936258 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nelfa | NM_011914.2 | 11 | 1676 | NP_036044.1 |
| AF101435.1 | 11 | 1628 | AAC72983.1 | |
| AK012673.1 | 1 | 245 | BAB28399.1 | |
| AK019191.1 | 11 | 1742 | ||
| AK078760.1 | 11 | 1676 | BAC37379.1 | |
| AK088384.1 | 11 | 1692 | BAC40319.1 | |
| AK146185.1 | 11 | 1728 | BAE26961.1 | |
| AK206019.1 | 1 | 149 | ||
| AK212506.1 | 1 | 85 | ||
| AK216954.1 | 1 | 171 | ||
| BC018423.1 | 2 | 289 | AAH18423.1 | |
| BC038003.1 | 11 | 1677 | AAH38003.1 |

Genomic Map