Assay Details
Target Gene Details
Entrez Gene ID: | 140494 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit A4 |
Gene Aliases: |
Atp6n1b, a4 |
Location: |
Chr.6:38048483-38124586 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v0a4 | NM_080467.3 | NP_536715.3 | ||
| AF326316.1 | AAL30435.1 | |||
| AF435090.1 | AAN45855.1 | |||
| AK128892.1 | BAC87655.1 | |||
| AK143790.1 | BAE25542.1 | |||
| AK165464.1 | BAE38201.1 | |||
| BC046979.1 | AAH46979.1 |
Target Gene Details
Entrez Gene ID: | 77522 |
Gene Name: | transmembrane protein 213 |
Gene Aliases: |
D630002J15Rik |
Location: |
Chr.6:38109353-38115806 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem213 | NM_029921.1 | 1 | 153 | NP_084197.1 |
| AK021320.1 | 1 | 154 | BAB32372.1 | |
| BC104374.1 | 1 | 145 | ||
| BC104375.1 | 1 | 145 | ||
| BC119477.1 | 1 | 145 |
More Information
Set Membership: |
|

Genomic Map