Assay Details
Target Gene Details
Entrez Gene ID: | 319776 |
Gene Name: | transmembrane protein 72 |
Gene Aliases: |
C230095G01Rik |
Location: |
Chr.6:116679103-116716978 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem72 | NM_178768.4 | 5 | 2394 | NP_848883.2 |
| AK049048.1 | 5 | 2549 | BAC33524.1 | |
| AK085603.1 | 5 | 2395 | BAC39484.1 | |
| AK141578.1 | 5 | 2551 | BAE24746.1 | |
| BC093486.1 | 5 | 2537 |

Genomic Map