Assay Details
Target Gene Details
Entrez Gene ID: | 23962 |
Gene Name: | 2'-5' oligoadenylate synthetase-like 2 |
Gene Aliases: |
M1204, Mmu-OASL, Oasl |
Location: |
Chr.5:114896934-114912245 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oasl2 | NM_011854.2 | 6 | 2681 | NP_035984.2 |
| AF068835.1 | 7 | 2612 | AAD02818.1 | |
| AK010034.1 | 6 | 2023 | BAB26655.1 | |
| AK150080.1 | 4 | 1472 | BAE29289.1 | |
| AK150492.1 | 6 | 2012 | BAE29608.1 | |
| AK160630.1 | 5 | 1585 | ||
| BC034361.1 | 6 | 2108 | AAH34361.1 |
More Information
Set Membership: |
|

Genomic Map