Assay Details
Target Gene Details
Entrez Gene ID: | 56715 |
Gene Name: | RAB guanine nucleotide exchange factor (GEF) 1 |
Gene Aliases: |
Rab5ef, Rabex5, Rin2 |
Location: |
Chr.5:130171791-130214342 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabgef1 | NM_001199059.1 | 8 | 1029 | NP_001185988.1 |
| NM_019983.2 | 7 | 1174 | NP_064367.1 | |
| AF093590.1 | 8 | 1005 | AAF28738.1 | |
| AK044568.1 | 7 | 1194 | BAC31984.1 | |
| AK146734.1 | 8 | 979 | BAE27396.1 | |
| BC018229.1 | 7 | 1174 | AAH18229.1 |
More Information
Set Membership: |
|

Genomic Map