Assay Details
Target Gene Details
Entrez Gene ID: | 19353 |
Gene Name: | RAS-related C3 botulinum substrate 1 |
Gene Aliases: |
AL023026, D5Ertd559e |
Location: |
Chr.5:143505481-143528116 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rac1 | NM_009007.2 | 6 | 1724 | NP_033033.1 |
| AK034601.1 | 6 | 1725 | BAC28767.1 | |
| AK047969.1 | 6 | 1727 | BAC33203.1 | |
| AK076023.1 | 6 | 1766 | BAC36128.1 | |
| AK081613.1 | 2 | 1253 | BAC38272.1 | |
| AK166383.1 | 7 | 1788 | BAE38744.1 | |
| BC003828.1 | 6 | 1725 | AAH03828.1 | |
| BC051053.1 | 6 | 1724 | AAH51053.1 | |
| BQ444803.1 | 1 | 486 | ||
| CX566679.1 | 1 | 220 |
More Information
Set Membership: |
|

Genomic Map