Assay Details
Target Gene Details
Entrez Gene ID: | 76223 |
Gene Name: | ATP/GTP binding protein-like 3 |
Gene Aliases: |
2900053G10Rik, 4930431N21Rik, 6530406M24Rik, CCP3 |
Location: |
Chr.6:34780432-34862184 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agbl3 | NM_001289656.1 | 8 | 1508 | NP_001276585.1 |
| NM_001289657.1 | 7 | 1482 | NP_001276586.1 | |
| NM_178630.4 | 7 | 1493 | NP_848745.2 | |
| AK029793.1 | 8 | 1509 | BAC26620.1 | |
| AK029798.1 | 7 | 1066 | ||
| AK030378.1 | 7 | 1051 | BAC26932.1 | |
| AK047649.1 | 7 | 1039 | BAC33111.1 | |
| AK054066.1 | 6 | 2513 | ||
| BC139065.1 | 7 | 945 | ||
| DQ867031.1 | 7 | 1494 | ||
| DQ867032.1 | 7 | 1494 |
More Information
Set Membership: |
|

Genomic Map