Assay Details
Target Gene Details
Entrez Gene ID: | 12344 |
Gene Name: | capping protein (actin filament) muscle Z-line, alpha 3 |
Gene Aliases: |
510-4, Cappa3, Gsg3, Tex8, repro32 |
Location: |
Chr.6:140041525-140042789 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Capza3 | NM_007605.4 | 1 | 208 | NP_031631.3 |
| AK016391.1 | 1 | 177 | BAB30213.1 | |
| BC049620.1 | 1 | 176 | AAH49620.1 | |
| BY707143.1 | 1 | 208 | ||
| D87471.1 | 1 | 166 | BAA13409.1 |
More Information
Set Membership: |
|

Genomic Map