Assay Details
Target Gene Details
Entrez Gene ID: | 56691 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily B, member 8 |
Gene Aliases: |
1700016F14Rik, 1700031F10Rik, AI326313, mDj6 |
Location: |
Chr.6:88222268-88223256 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajb8 | NM_019964.1 | 1 | 164 | NP_064348.1 |
| AB028856.1 | 1 | 164 | BAA88304.1 | |
| AK006026.1 | 1 | 148 | BAB24372.1 | |
| BC049591.1 | 1 | 142 | AAH49591.1 | |
| BC061112.1 | 1 | 147 | AAH61112.1 |

Genomic Map