Assay Details
Target Gene Details
Entrez Gene ID: | 317758 |
Gene Name: | GTPase, IMAP family member 9 |
Gene Aliases: |
A630002K24, BB145400 |
Location: |
Chr.6:48676121-48679116 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gimap9 | NM_174960.2 | 2 | 334 | NP_777620.1 |
| AK041336.1 | 2 | 323 | BAC30911.1 | |
| BB618178.1 | 2 | 334 | ||
| BC038043.1 | 2 | 256 | AAH38043.1 | |
| BC096680.1 | 2 | 318 | AAH96680.1 |
More Information
Set Membership: |
|

Genomic Map