Assay Details
Target Gene Details
Entrez Gene ID: | 231130 |
Gene Name: | TNFAIP3 interacting protein 2 |
Gene Aliases: |
1810020H16Rik, ABIN-2, AI428870 |
Location: |
Chr.5:34496096-34513979 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnip2 | NM_139064.2 | 6 | 1258 | NP_620703.1 |
| AJ304865.1 | 6 | 1258 | CAC34841.1 | |
| AK160165.1 | 6 | 1216 | BAE35666.1 | |
| BC052083.1 | 7 | 1304 | AAH52083.1 |

Genomic Map