Assay Details
Target Gene Details
Entrez Gene ID: | 11997 |
Gene Name: | aldo-keto reductase family 1, member B7 |
Gene Aliases: |
AR, Avdp, MVDP |
Location: |
Chr.6:34412362-34423137 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1b7 | NM_009731.2 | 10 | 1148 | NP_033861.2 |
| AK002705.1 | 10 | 1150 | BAB22299.1 | |
| AK147775.1 | 10 | 1177 | BAE28129.1 | |
| BC086929.1 | 10 | 1121 | AAH86929.1 | |
| J05663.1 | 10 | 1120 | AAA39773.1 |
More Information
Set Membership: |
|

Genomic Map