Assay Details
Target Gene Details
Entrez Gene ID: | 15398 |
Gene Name: | homeobox A13 |
Gene Aliases: |
Hd, Hox-1.10 |
Location: |
Chr.6:52258853-52260880 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxa13 | NM_008264.1 | 2 | 1224 | NP_032290.1 |
| BC100731.1 | 3 | 693 | AAI00732.1 | |
| BC100732.1 | 3 | 690 | AAI00733.1 | |
| BC100733.1 | 3 | 690 | AAI00734.1 |

Genomic Map