Assay Details
Target Gene Details
Entrez Gene ID: | 319734 |
Gene Name: | calcium channel, voltage-dependent, alpha 2/delta subunit 4 |
Gene Aliases: |
5730412N02Rik, BE686333 |
Location: |
Chr.6:119236474-119353392 on Build GRCm38 |
Assay Gene Location: | Within Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacna2d4 | NM_001033382.2 | NP_001028554.2 | ||
| BC141091.1 |
Target Gene Details
Entrez Gene ID: | 211187 |
Gene Name: | leucine-rich repeats and transmembrane domains 2 |
Gene Aliases: |
A230084J22, AI841794 |
Location: |
Chr.6:119315133-119332270 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrtm2 | NM_001172207.1 | 5 | 2615 | NP_001165678.1 |
| NM_172492.3 | 5 | 2786 | NP_766080.1 | |
| AK039006.1 | 5 | 2620 | BAC30200.1 | |
| AK045494.1 | 5 | 2700 | BAC32394.1 | |
| BC058944.1 | 4 | 3176 | AAH58944.1 |
More Information
Set Membership: |
|

Genomic Map