Assay Details
Target Gene Details
Entrez Gene ID: | 13619 |
Gene Name: | polyhomeotic-like 1 (Drosophila) |
Gene Aliases: |
AW557034, Edr, Edr1, Mph1, Rae-28, rae28 |
Location: |
Chr.6:122317731-122340227 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phc1 | NM_001042623.2 | 14 | 3295 | NP_001036088.1 |
| NM_001271579.1 | 15 | 3422 | NP_001258508.1 | |
| NM_007905.3 | 15 | 3451 | NP_031931.2 | |
| NR_073364.1 | 15 | 3458 | ||
| BC046535.1 | 15 | 3438 | AAH46535.1 | |
| BC052394.1 | 15 | 3784 | AAH52394.1 | |
| U63386.1 | 14 | 3379 | AAC28974.1 |
More Information
Set Membership: |
|

Genomic Map