Assay Details
Target Gene Details
Entrez Gene ID: | 50926 |
Gene Name: | heterogeneous nuclear ribonucleoprotein D-like |
Gene Aliases: |
AA407431, AA959857, D5Ertd650e, D5Wsu145e, Hnrnpdl1, Hnrpdl, JKTBP, hnRNP DL, hnRNP-DL |
Location: |
Chr.5:100033577-100039950 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpdl | NM_016690.4 | 1 | 90 | NP_057899.2 |
| BM950445.1 | 1 | 90 |

Genomic Map