Assay Details
Target Gene Details
Entrez Gene ID: | 67054 |
Gene Name: | phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoribosylaminoimidazole, succinocarboxamide synthetase |
Gene Aliases: |
2610511I09Rik, ADE2H1, AIRC, PAIS |
Location: |
Chr.5:76951315-76967507 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Paics | NM_025939.2 | 1 | 286 | NP_080215.1 |
| AK012118.1 | 1 | 44 | BAB28044.1 | |
| AK146722.1 | BAE27385.1 | |||
| AK159398.1 | 1 | 90 | BAE35051.1 | |
| BC052691.1 | 1 | 286 | AAH52691.1 |

Genomic Map