Assay Details
Target Gene Details
Entrez Gene ID: | 56522 |
Gene Name: | poly (A) polymerase beta (testis specific) |
Gene Aliases: |
PAP, Paplob, Papola-ps, Papt, Plap-ps, Tpap |
Location: |
Chr.5:142527740-142530076 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Papolb | NM_019943.2 | 1 | 1949 | NP_064327.1 |
| AB030729.1 | 1 | 1949 | BAA83086.1 | |
| AB221598.1 | 1 | 1759 | ||
| AF039957.1 | 1 | 1899 | AAD43624.1 | |
| BC129896.1 | 1 | 1927 | ||
| BC129897.1 | 1 | 1927 |
Target Gene Details
Entrez Gene ID: | 231858 |
Gene Name: | Ras association and DIL domains |
Gene Aliases: |
AI536456, D930005D10Rik |
Location: |
Chr.5:142484839-142551098 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Radil | NM_001289588.1 | NP_001276517.1 | ||
| NM_178702.4 | NP_848817.2 | |||
| AK038570.1 | BAC30049.1 | |||
| AK052964.1 | BAC35223.1 | |||
| AK083410.1 | BAC38904.1 | |||
| AK154169.1 | ||||
| AK162917.1 | ||||
| AK163852.1 | ||||
| AK173277.1 | BAD32555.1 | |||
| BC056483.1 | AAH56483.1 |
More Information
Set Membership: |
|

Genomic Map