Assay Details
Target Gene Details
Entrez Gene ID: | 215160 |
Gene Name: | rhomboid domain containing 2 |
Gene Aliases: |
0610011L13Rik, AI480532, Rhbdl7, Usmg1 |
Location: |
Chr.5:135632654-135646376 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhbdd2 | NM_146002.2 | 4 | 3601 | NP_666114.1 |
| AK032400.1 | 3 | 3025 | ||
| AK141182.1 | 5 | 3722 |
More Information
Set Membership: |
|

Genomic Map