Assay Details
Target Gene Details
Entrez Gene ID: | 20599 |
Gene Name: | submaxillary gland androgen regulated protein 3A |
Gene Aliases: |
AA589601, MSG1, MSG3, Smr1, Smr3 |
Location: |
Chr.5:87969504-88008534 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smr3a | NM_011422.3 | 3 | 412 | NP_035552.1 |
| AA727574.1 | ||||
| BC031806.1 | 3 | 378 | AAH31806.1 | |
| BC031921.1 | 3 | 356 | AAH31921.1 | |
| BC034553.1 | 3 | 382 | AAH34553.1 | |
| BC055857.1 | 3 | 406 | AAH55857.1 | |
| BC141243.1 | 3 | 374 | ||
| BC145569.1 | 3 | 374 | ||
| BF454619.1 | 3 | 380 | ||
| X71629.1 | 3 | 389 | CAA50636.1 |

Genomic Map