Assay Details
Target Gene Details
Entrez Gene ID: | 76167 |
Gene Name: | small nuclear ribonucleoprotein 35 (U11/U12) |
Gene Aliases: |
6330548G22Rik |
Location: |
Chr.5:124483080-124491323 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snrnp35 | NM_029532.2 | 2 | 262 | NP_083808.1 |
| AK018232.1 | 2 | 237 | BAB31127.1 | |
| AK039064.1 | 2 | 284 | BAC30228.1 | |
| BC043031.1 | 2 | 262 | AAH43031.1 | |
| BC058361.1 | 2 | 262 | AAH58361.1 |
More Information
Set Membership: |
|

Genomic Map