Assay Details
Target Gene Details
Entrez Gene ID: | 22051 |
Gene Name: | thyroid hormone receptor interactor 6 |
Gene Aliases: |
- |
Location: |
Chr.5:137309899-137314487 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trip6 | NM_011639.3 | 4 | 863 | NP_035769.1 |
| AF097511.1 | 4 | 756 | AAC72380.1 | |
| AF116823.1 | 4 | 858 | AAD45984.1 | |
| AK010435.1 | 4 | 864 | BAB26937.1 | |
| AK146823.1 | 4 | 812 | ||
| AK159387.1 | 4 | 832 | BAE35042.1 | |
| AK166024.1 | 4 | 1027 | BAE38527.1 | |
| AK215713.1 | 1 | 295 | ||
| BC138685.1 | 4 | 795 | ||
| BC138686.1 | 4 | 795 |
More Information
Set Membership: |
|

Genomic Map