Assay Details
Target Gene Details
Entrez Gene ID: | 11844 |
Gene Name: | ADP-ribosylation factor 5 |
Gene Aliases: |
- |
Location: |
Chr.6:28423640-28426499 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arf5 | NM_007480.1 | 6 | 719 | NP_031506.1 |
| BC132452.1 | 7 | 707 | ||
| BC138756.1 | 7 | 707 | ||
| D87902.1 | 6 | 719 | BAA13494.1 |
More Information
Set Membership: |
|

Genomic Map