Assay Details
Target Gene Details
Entrez Gene ID: | 667992 |
Gene Name: | taste receptor, type 2, member 103 |
Gene Aliases: |
EG667992, T2R3, TRB2, Tas2r10, Tas2r3, mGR03, mt2r63 |
Location: |
Chr.6:133036163-133037101 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas2r103 | NM_053211.1 | 1 | 206 | NP_444441.1 |
More Information
Set Membership: |
|

Genomic Map