Assay Details
Target Gene Details
Entrez Gene ID: | 15874 |
Gene Name: | islet amyloid polypeptide |
Gene Aliases: |
DAP |
Location: |
Chr.6:142298425-142303820 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iapp | NM_010491.2 | 3 | 269 | NP_034621.1 |
| BC027527.1 | 3 | 240 | AAH27527.1 | |
| CJ042995.1 | 3 | 272 | ||
| M25389.1 | 2 | 176 | AAA37874.1 |
Target Gene Details
Entrez Gene ID: | 108096 |
Gene Name: | solute carrier organic anion transporter family, member 1a5 |
Gene Aliases: |
Oatp-3, Oatp3, Slc21a7 |
Location: |
Chr.6:142234227-142322981 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slco1a5 | NM_130861.3 | NP_570931.1 | ||
| AF240694.1 | AAK39416.1 | |||
| AK029706.1 |
More Information
Set Membership: |
|

Genomic Map