Assay Details
Target Gene Details
Entrez Gene ID: | 17120 |
Gene Name: | MAD1 mitotic arrest deficient 1-like 1 |
Gene Aliases: |
AI173502, AW550425, Mad1 |
Location: |
Chr.5:140008689-140321591 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mad1l1 | NM_010752.3 | 18 | 2158 | NP_034882.2 |
| AF083812.1 | 18 | 2165 | AAD24499.1 | |
| AF083813.1 | 17 | 2074 | AAD24500.1 | |
| AK136981.1 | 18 | 2161 | ||
| AK169976.1 | 7 | 1127 | BAE41492.1 | |
| BC010702.1 | 18 | 2156 | AAH10702.1 |

Genomic Map