Assay Details
Target Gene Details
Entrez Gene ID: | 18542 |
Gene Name: | procollagen C-endopeptidase enhancer protein |
Gene Aliases: |
Astt-2, Astt2, P14, PCPE-1 |
Location: |
Chr.5:137605105-137611899 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcolce | NM_008788.2 | 3 | 449 | NP_032814.2 |
| AB008548.1 | 3 | 373 | BAA23280.1 | |
| AK142615.1 | 3 | 378 | BAE25130.1 | |
| AK144392.1 | 3 | 450 | BAE25865.1 | |
| AK146829.1 | 3 | 450 | BAE27464.1 | |
| AK159430.1 | 3 | 437 | BAE35077.1 | |
| AK159940.1 | 3 | 442 | ||
| AK162524.1 | 3 | 453 | ||
| CX210896.1 | 1 | 131 | ||
| X57337.1 | 3 | 416 | CAA40612.1 |
More Information
Set Membership: |
|

Genomic Map