Assay Details
Target Gene Details
Entrez Gene ID: | 74570 |
Gene Name: | zinc finger with KRAB and SCAN domains 1 |
Gene Aliases: |
5930429A01Rik, 9130423L19Rik, 9230118B16Rik, AI646829, AI788588, C87323, KOX18, PHZ-37, ZNF139 |
Location: |
Chr.5:138085076-138107822 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zkscan1 | NM_029869.1 | 4 | 6826 | NP_084145.1 |
| NM_133906.4 | 6 | 7051 | NP_598667.2 | |
| AK033176.1 | 1 | 2329 | BAC28182.1 |
More Information
Set Membership: |
|

Genomic Map