Assay Details
Target Gene Details
Entrez Gene ID: | 54324 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 5 |
Gene Aliases: |
2210412D05Rik, AA717842, AW495314, Tim1 |
Location: |
Chr.6:43265644-43289320 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgef5 | NM_133674.1 | 15 | 4820 | NP_598435.1 |
| AK020295.1 | 3 | 584 | ||
| AK220242.1 | 4 | 1737 | BAD90167.1 | |
| BC006829.1 | 9 | 2515 | AAH06829.1 | |
| BC025127.1 | 10 | 1107 | AAH25127.1 | |
| CB058042.1 | 3 | 298 |
More Information
Set Membership: |
|

Genomic Map