Assay Details
Target Gene Details
Entrez Gene ID: | 72772 |
Gene Name: | RAD50 interactor 1 |
Gene Aliases: |
1500019C06Rik, 2810450M21Rik, Rint-1 |
Location: |
Chr.5:23787681-23820369 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rint1 | NM_177323.3 | 15 | 2939 | NP_796297.2 |
| AK028707.1 | 14 | 2796 | BAC26077.1 | |
| AK133789.1 | 15 | 3133 | ||
| AK144144.1 | 15 | 2962 | ||
| BC065059.1 | 15 | 2939 | AAH65059.2 |

Genomic Map