Assay Details
Target Gene Details
Entrez Gene ID: | 109267 |
Gene Name: | scavenger receptor cysteine rich family, 4 domains |
Gene Aliases: |
C330016E03Rik, S4D-SRCRB, SRCRB-S4D, Srcrb4d |
Location: |
Chr.5:135960220-135974535 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssc4d | NM_001160366.1 | 11 | 2235 | NP_001153838.1 |
| AI326785.1 | 1 | 494 | ||
| AK082786.1 | 3 | 646 | BAC38619.1 | |
| BC117747.1 | ||||
| BC126947.1 |
More Information
Set Membership: |
|

Genomic Map