Assay Details
Target Gene Details
Entrez Gene ID: | 109552 |
Gene Name: | sorcin |
Gene Aliases: |
2210417O06Rik, 2900070H08Rik, Sor |
Location: |
Chr.5:8046078-8069314 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sri | NM_001080974.2 | 8 | 1187 | NP_001074443.1 |
| NM_025618.3 | 8 | 1227 | NP_079894.2 | |
| AK134747.1 | 8 | 1189 | BAE22266.1 | |
| AK146911.1 | 8 | 1168 | BAE27523.1 | |
| AK168572.1 | 8 | 1168 | BAE40442.1 | |
| AK168900.1 | 8 | 1159 | BAE40716.1 |
More Information
Set Membership: |
|

Genomic Map