Assay Details
Target Gene Details
Entrez Gene ID: | 65255 |
Gene Name: | ankyrin repeat and SOCS box-containing 4 |
Gene Aliases: |
8430401O13Rik, AV117194, AW060255 |
Location: |
Chr.6:5383386-5433022 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asb4 | NM_023048.5 | 4 | 757 | NP_075535.1 |
| AF155355.1 | 3 | 727 | AAD38811.1 | |
| AK018373.1 | 3 | 910 | BAB31183.1 | |
| AK076004.1 | 3 | 814 | ||
| AK133400.1 | 2 | 360 | BAE21634.1 | |
| AK133894.1 | 3 | 628 | BAE21916.1 | |
| BC046819.1 | 4 | 757 | AAH46819.1 | |
| BC054745.1 | 3 | 865 | AAH54745.1 | |
| BC056440.1 | 4 | 757 | AAH56440.1 |
More Information
Set Membership: |
|

Genomic Map