Assay Details
Target Gene Details
Entrez Gene ID: | 22226 |
Gene Name: | urocortin |
Gene Aliases: |
Mpv17, Ucn1 |
Location: |
Chr.5:31137989-31138895 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ucn | NM_021290.2 | 2 | 468 | NP_067265.1 |
| BC116948.1 | 2 | 402 | ||
| BC116950.1 | 2 | 402 | ||
| BC144891.1 | 2 | 397 |
More Information
Set Membership: |
|

Genomic Map