Assay Details
Target Gene Details
Entrez Gene ID: | 243764 |
Gene Name: | cholinergic receptor, muscarinic 2, cardiac |
Gene Aliases: |
AChR-M2, Chrm-2, M2 |
Location: |
Chr.6:36387950-36528638 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrm2 | NM_203491.3 | 3 | 1407 | NP_987076.2 |
More Information
Set Membership: |
|

Genomic Map