Assay Details
Target Gene Details
Entrez Gene ID: | 12723 |
Gene Name: | chloride channel 1 |
Gene Aliases: |
Clc-1, Clc1, SMCC1, adr, mto, myotonia, nmf355 |
Location: |
Chr.6:42282593-42315759 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clcn1 | NM_013491.2 | 1 | 211 | NP_038519.1 |
| AK036735.1 | 1 | 213 | BAC29557.1 | |
| AY046403.1 | AAL05907.1 | |||
| AY046404.1 | AAL05908.1 | |||
| BC114336.1 | 1 | 176 |
More Information
Set Membership: |
|

Genomic Map