Assay Details
Target Gene Details
Entrez Gene ID: | 12723 |
Gene Name: | chloride channel 1 |
Gene Aliases: |
Clc-1, Clc1, SMCC1, adr, mto, myotonia, nmf355 |
Location: |
Chr.6:42282593-42315759 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clcn1 | NM_013491.2 | 23 | 3401 | NP_038519.1 |
| AJ011107.1 | 1 | 334 | ||
| AK036735.1 | 23 | 3403 | BAC29557.1 | |
| AK161489.1 | 15 | 2261 | BAE36420.1 |
More Information
Set Membership: |
|

Genomic Map