Assay Details
Target Gene Details
Entrez Gene ID: | 56524 |
Gene Name: | membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6) |
Gene Aliases: |
P55t, Pals2 |
Location: |
Chr.6:50110241-50198598 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpp6 | NM_001164733.1 | 14 | 1908 | NP_001158205.1 |
| NM_001164734.1 | 14 | 1918 | NP_001158206.1 | |
| NM_019939.2 | 13 | 1876 | NP_064323.2 | |
| AF161181.1 | 13 | 1740 | AAD45009.1 | |
| AF199009.1 | 13 | 1872 | AAF63790.1 | |
| AF199010.1 | 14 | 1914 | AAF63791.1 | |
| AK077754.1 | 4 | 494 | BAC36993.1 | |
| AK144434.1 | 14 | 1919 | BAE25887.1 | |
| BC138666.1 | 13 | 1734 | ||
| BC145363.1 | 14 | 1776 | ||
| BC145364.1 | 14 | 1766 |
More Information
Set Membership: |
|

Genomic Map