Assay Details
Target Gene Details
Entrez Gene ID: | 58182 |
Gene Name: | prokineticin receptor 1 |
Gene Aliases: |
EG-VEGFR1, Gpr73, Pkr1 |
Location: |
Chr.6:87578589-87590774 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prokr1 | NM_021381.3 | 2 | 527 | NP_067356.2 |
| AF236082.1 | 1 | 281 | AAF43706.1 | |
| AF487278.1 | 1 | 281 | AAM49570.1 | |
| AK134279.1 | 2 | 541 | BAE22080.1 | |
| AK156734.1 | 2 | 701 | BAE33828.1 | |
| BC059003.1 | 2 | 527 | AAH59003.1 |
More Information
Set Membership: |
|

Genomic Map