Assay Details
Target Gene Details
Entrez Gene ID: | 100042056 |
Gene Name: | RIKEN cDNA 9130019P16 gene |
Gene Aliases: |
6430501H15Rik, OTTMUSG00000023151 |
Location: |
Chr.6:54269681-54430221 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9130019P16Rik | NR_033635.1 | |||
| AK032212.1 | BAC27762.1 |
Target Gene Details
Entrez Gene ID: | 78004 |
Gene Name: | proline rich 15 |
Gene Aliases: |
E130201N16Rik, G90 |
Location: |
Chr.6:54327012-54330200 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prr15 | NM_030024.2 | 2 | 480 | NP_084300.1 |
| AJ132433.2 | 2 | 480 | CAD45579.1 | |
| AK021401.1 | 2 | 588 | BAB32398.1 | |
| AK136438.1 | 2 | 657 | ||
| BC132416.1 | 1 | 156 | ||
| BC138757.1 | 1 | 156 |
More Information
Set Membership: |
|

Genomic Map